Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, for 8mm OD Standard Wall or 9mm OD Medium Wall, Red
SDP
Supplier:  Chemglass Life Sciences CG302295
Encompass_Preferred
Septum Stopper, Sleeve Type, for 8mm OD Standard Wall or 9mm OD Medium Wall, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. 50-872-550