Learn More
Chemglass Life Sciences Septum Stopper, Suba-Seal, For 29/26-29/42 Outer Joints and 25.5mm ID Tubing
SDP
Supplier:  Chemglass Life Sciences CG302409
Encompass_Preferred
Septum Stopper, Suba-Seal, For 29/26-29/42 Outer Joints and 25.5mm ID Tubing. Suba-Seal septa are a flanged stopper with a hollow plug and sleeve extension that folds down over the neck of the vessel. In addition to the pull over sleeve, Suba-Seal stoppers have a series of annular serrations on the plug portion of the septa. Each of these serrations depress along the inner wall of the glass vessel to form an extremely effective air-tight seal. Stoppers are made from natural red rubber and sold in the package quantities listed below.
Catalog No. 50-872-554