Learn More
Cayman Chemical SpUrIdIn5O 1thIotrIpho 2.5mg
SDP
Catalog No. 502919123
Encompass_Preferred
A nucleotide P2Y2 and P2Y4 receptor agonist; induces inositol phosphate accumulation in 1321N1 cells expressing the purinergic P2Y2 or P2Y4 receptors (EC50s = 14 and 81 µM, respectively)
Catalog No. 50-291-9123 Supplier Cayman Chemical Supplier No. 385732.5 MG
A nucleotide P2Y2 and P2Y4 receptor agonist; induces inositol phosphate accumulation in 1321N1 cells expressing the purinergic P2Y2 or P2Y4 receptors (EC50s = 14 and 81 µM, respectively)