Learn More
Sigma Aldrich Fine Chemicals Biosciences Spurr Low Viscosity Embedd
Catalog No. 502904484
Encompass
Spurr resin is a hydrophobic resin. It is less viscous than epon mixtures and is easier to dispense. The low viscosity of the resin contributes to its penetration in tissue mineral and dense structures. A low viscosity of 60 cps also allows easy penetration into a variety of difficult materials.
Catalog No. 50-290-4484 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. EM03001KT
Spurr resin is a hydrophobic resin. It is less viscous than epon mixtures and is easier to dispense. The low viscosity of the resin contributes to its penetration in tissue mineral and dense structures. A low viscosity of 60 cps also allows easy penetration into a variety of difficult materials.