Learn More
Polysciences, Inc. Spurr Low Viscosity Embedding Kit
SDP
Catalog No. NC0607864
Encompass
Spurr Low Viscosity Embedding kit provides exceptional penetration in tissue, mineral, and dense structures. A low viscosity of 60cps allows easy penetration into a variety of difficult materials. May be used to prepare mineral specimens for polishing. The low viscosity of the Spurr formulation allows rapid infiltration into tissues, minerals and other dense structures. Facilitates embedments with high lipid contents or hard membranes or highly vacuolated parenchymatous tissues.
Catalog No. NC0607864 Supplier Polysciences, Inc. Supplier No. 019161
Spurr Low Viscosity Embedding kit provides exceptional penetration in tissue, mineral, and dense structures. A low viscosity of 60cps allows easy penetration into a variety of difficult materials. May be used to prepare mineral specimens for polishing. The low viscosity of the Spurr formulation allows rapid infiltration into tissues, minerals and other dense structures. Facilitates embedments with high lipid contents or hard membranes or highly vacuolated parenchymatous tissues.