Learn More
Electron Microscopy Sciences Sputtering Target Tantalum, 50.8 mm
SDP
Catalog No. 501901438
Encompass_Preferred
EMS precious metal sputtering targets are made from high purity metals. The targets are for use on most sputtering units and includes systems manufacturers Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and others. Our targets come in a standard thickness of 0.1 mm (100um). Targets are available mounted or unmounted. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you.
EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.
Diameter: 50.8 mm
Catalog No. 50-190-1438 Supplier Electron Microscopy Sciences Supplier No. 91015TA
EMS precious metal sputtering targets are made from high purity metals. The targets are for use on most sputtering units and includes systems manufacturers Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and others. Our targets come in a standard thickness of 0.1 mm (100um). Targets are available mounted or unmounted. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you.
EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.
Diameter: 50.8 mm