Learn More
ADHESIVE TAPE PRODUCTS LTD SPVC EL TAPE BL 2'X66' 80R
Catalog No. NC9641345
Encompass_Preferred
7mil (0.178mm) PVC Electrical Insulation Tape, Natural Rubber adhesive. For wire and cable insulation, bundling, primary splice insulation at no more than 600V. Non-flame retardant.
Catalog No. NC9641345 Supplier ADHESIVE TAPE PRODUCTS LTD Supplier No. EL766B
7mil (0.178mm) PVC Electrical Insulation Tape, Natural Rubber adhesive. For wire and cable insulation, bundling, primary splice insulation at no more than 600V. Non-flame retardant.