Learn More
Cytoskeleton, Inc. SPY555-DNA Fluorogenic DNA Stain, Bright Non-Toxic, Low Background, 1 vial
Catalog No. NC2340163
Encompass
SPY555-DNA is a bright, non-toxic live cell nuclear stain that enables quick and specific DNA labeling in live and fixed cells. It features very low background and is optimized for high sensitivity, making it suitable for advanced imaging techniques like confocal and STED microscopy. Its absorbance and emission properties are similar to TMR, allowing compatibility with standard filter sets, which enhances multicolor imaging alongside other dyes.
Catalog No. NC2340163 Supplier Cytoskeleton, Inc. Supplier No. CYSC201
SPY555-DNA is a bright, non-toxic live cell nuclear stain that enables quick and specific DNA labeling in live and fixed cells. It features very low background and is optimized for high sensitivity, making it suitable for advanced imaging techniques like confocal and STED microscopy. Its absorbance and emission properties are similar to TMR, allowing compatibility with standard filter sets, which enhances multicolor imaging alongside other dyes.