Learn More
Cytoskeleton, Inc. SPY555-FastAct™ F-Actin Stain, Non-Toxic, Compatible With Live Cell Imaging
Catalog No. NC2604417
Encompass
The SPY555-FastAct™ from Cytoskeleton, Inc. is an innovative orange F-actin stain that enables high-specificity, low-background labeling of F-actin in live cells. Optimized for fast actin dynamics, it requires no genetic manipulation, making it ideal for multicolor imaging alongside other SPY™ dyes or GFP. It is lyophilized for convenience, reconstituting to create a stable 1000x stock solution for 100 stainings under recommended conditions. Perfect for advanced microscopy techniques.
Catalog No. NC2604417 Supplier Cytoskeleton, Inc. Supplier No. CYSC205
The SPY555-FastAct™ from Cytoskeleton, Inc. is an innovative orange F-actin stain that enables high-specificity, low-background labeling of F-actin in live cells. Optimized for fast actin dynamics, it requires no genetic manipulation, making it ideal for multicolor imaging alongside other SPY™ dyes or GFP. It is lyophilized for convenience, reconstituting to create a stable 1000x stock solution for 100 stainings under recommended conditions. Perfect for advanced microscopy techniques.