Learn More
Cytoskeleton, Inc. SPY555-Tubulin Live Cell Stain, Bright Fluorogenic Probe, Compatible with Advanced Microscopy Techniques, 1 vial
Catalog No. NC2856127
Encompass
SPY555-tubulin is a non-toxic live-cell probe designed for specific and low-background fluorescence imaging of microtubules. This product enables rapid microtubule labeling in living cells without genetic manipulation or fluorescent protein overexpression. It supports advanced microscopy techniques and multicolor imaging, providing flexibility and ease of use in various imaging applications.
Catalog No. NC2856127 Supplier Cytoskeleton, Inc. Supplier No. CYSC203
Add to Cart
Add to Cart
Request a Quote
SPY555-tubulin is a non-toxic live-cell probe designed for specific and low-background fluorescence imaging of microtubules. This product enables rapid microtubule labeling in living cells without genetic manipulation or fluorescent protein overexpression. It supports advanced microscopy techniques and multicolor imaging, providing flexibility and ease of use in various imaging applications.