Learn More
Cytoskeleton, Inc. SPY650-DNA Kit, Live Cell Nuclear Stain, Far-Red Fluorescence, 100 Stainings
Catalog No. NC2096299
Encompass
The SPY650-DNA Kit from Cytoskeleton, Inc. is a highly specific live cell nuclear stain designed for DNA labeling in live and fixed cells. Its far-red fluorescent properties reduce phototoxicity and autofluorescence, making it ideal for advanced imaging techniques like confocal and STED microscopy. Each kit provides sufficient reagent for 100 stainings, facilitating multicolor imaging without genetic manipulation.
This item is not returnable. View return policy
Catalog No. NC2096299 Supplier Cytoskeleton, Inc. Supplier No. CYSC501
The SPY650-DNA Kit from Cytoskeleton, Inc. is a highly specific live cell nuclear stain designed for DNA labeling in live and fixed cells. Its far-red fluorescent properties reduce phototoxicity and autofluorescence, making it ideal for advanced imaging techniques like confocal and STED microscopy. Each kit provides sufficient reagent for 100 stainings, facilitating multicolor imaging without genetic manipulation.