Learn More
Cytoskeleton, Inc. SPY650-FastAct™, Non-toxic Live Cell F-actin Stain, Compatible with Widefield & Confocal Microscopy, 1 vial
Supplier:  Cytoskeleton, Inc. CYSC505
Encompass
The SPY650-FastAct™ is a non-toxic, far-red fluorogenic probe for live cell F-actin staining, enabling high specificity and low background in labeling dynamic and static actin filaments. This product differentiates itself by providing visualization of fast actin dynamics without genetic manipulation, utilizing the superior photostable SPY650 fluorophore compatible with standard imaging techniques. Ideal for researchers needing precise imaging in live cell contexts.
Catalog No. NC2204057