Learn More
Aldevron SpyFi Cas9 Nuclease, 0.25mg
Catalog No. 503073894
Encompass_Preferred
0.25mg dispense of SpyFi Cas9 Nuclease. From basic research to clinical applications, our SpyFi Cas9 Nuclease delivers greater editing efficiency and reduced off-target effects without loss of on-target activity to help accelerate your CRISPR-Cas9 gene editing program. SpyFi Cas9 Nucleases are available at Research-Grade through Fisher, or across all quality grades (Research Grade or GMP) directly through Aldevron.
SpyFi Cas9 Nuclease is uniquely positioned to satisfy a wide range of needs from basic research to clinical applications. When used in combination with effective guide, target site and double-stranded DNA cut strategies, SpyFi Cas9 Nuclease delivers greater editing efficiency, higher fidelity and less off-target activity than wild-type SpCas9 nuclease.
Functional Performance
•Reduced off-target effects
•Ready to use for electroporation or transfection
•10 mg/mL formulation for ease of use
Catalog No. 50-307-3894 Supplier Aldevron Supplier No. 92140.25MG
0.25mg dispense of SpyFi Cas9 Nuclease. From basic research to clinical applications, our SpyFi Cas9 Nuclease delivers greater editing efficiency and reduced off-target effects without loss of on-target activity to help accelerate your CRISPR-Cas9 gene editing program. SpyFi Cas9 Nucleases are available at Research-Grade through Fisher, or across all quality grades (Research Grade or GMP) directly through Aldevron.
SpyFi Cas9 Nuclease is uniquely positioned to satisfy a wide range of needs from basic research to clinical applications. When used in combination with effective guide, target site and double-stranded DNA cut strategies, SpyFi Cas9 Nuclease delivers greater editing efficiency, higher fidelity and less off-target activity than wild-type SpCas9 nuclease.
Functional Performance
•Reduced off-target effects
•Ready to use for electroporation or transfection
•10 mg/mL formulation for ease of use