Learn More
Aviva Systems Biology Corp SPZ1 Antibody - C-terminal region (ARP37405_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37405_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SPZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse SPZ1. Alternate names: AI429130, 1700027M20RikApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53072