Learn More
Enzo Life Sciences SQ 22536 (5mg). CAS: 17318-31-9
SDP
Supplier:  Enzo Life Sciences BMLCN1400005
Encompass_Preferred
SQ 22536 is a cell-permeable adenylate cyclase inhibitor. It inhibits cAMP production induced by prostaglandin E1 in platelet lysates (IC50=82 µM) and in intact platelets (IC50=1 µM). It has been successfully employed as a probe for the involvement of adenylate cyclase in a variety of receptor systems including those for prostaglandins and neurotransmitters. ReducesPGE2-induced inhibition of O2- production. Alternative name: 9-THF-Ade. Purity: ≥99% (TLC). Solubility: Soluble in water (25mg/ml) or DMSO (50mM with warming). Long Term Storage: -20°C.
Catalog No. 50-201-0445