Learn More
Malvern Panalytical Square Cell with Cap (12mm O.D.)
Catalog No. NC0631452
Encompass
2mm o.d. square glass cell for size measurements. For: the Zetasizer Nano & Zetasizer Advanec series and Zetasizer V.With circular aperture and stopper. Not suitable for use with the 'dip' electrophoresis electrodes
Catalog No. NC0631452 Supplier Malvern Panalytical Supplier No. 10049486
2mm o.d. square glass cell for size measurements. For: the Zetasizer Nano & Zetasizer Advanec series and Zetasizer V.With circular aperture and stopper. Not suitable for use with the 'dip' electrophoresis electrodes