Learn More
Electron Microscopy Sciences Veco Square Center Reference 150 mesh Copper Grid
SDP
Catalog No. 50980111
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm
Catalog No. 50-980-111 Supplier Electron Microscopy Sciences Supplier No. 0150CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm