Learn More
Electron Microscopy Sciences Veco Square Center Reference 50 mesh Nickel Grid
SDP
Catalog No. 50980102
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 500 µm
Hole: 450 µm
Bar: 50 µm
Catalog No. 50-980-102 Supplier Electron Microscopy Sciences Supplier No. 0050NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 500 µm
Hole: 450 µm
Bar: 50 µm