Learn More
Electron Microscopy Sciences Veco Square Center Reference 100 mesh Gold Grid
SDP
Catalog No. 50980106
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm
Catalog No. 50-980-106 Supplier Electron Microscopy Sciences Supplier No. 0100AU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm