Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Veco Square Center Reference 100 mesh Copper Grid
SDP

Supplier:  Electron Microscopy Sciences 0100CU

Encompass_Preferred

Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 250 µm
Hole: 200 µm
Bar: 50 µm

Catalog No. 50-980-108


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.