Learn More
Electron Microscopy Sciences Veco Square Center Reference 400 mesh Gold Grid
SDP
Catalog No. 50980125
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm
Catalog No. 50-980-125 Supplier Electron Microscopy Sciences Supplier No. 0400AU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm