Learn More
Electron Microscopy Sciences Veco Square Center Reference 400 mesh Copper Grid
SDP
Catalog No. 50980126
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm
Catalog No. 50-980-126 Supplier Electron Microscopy Sciences Supplier No. 0400CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm