Learn More
Electron Microscopy Sciences Veco Square Center Reference 200 mesh Grid, Stainless
SDP
Catalog No. 50980117
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter. Punched from Stainless Steel Woven 200 mesh.
Grid: Stainless
Catalog No. 50-980-117 Supplier Electron Microscopy Sciences Supplier No. 0200SS
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter. Punched from Stainless Steel Woven 200 mesh.
Grid: Stainless