Learn More
Grainger Sq. End Mill Single End Carb 0.0180
Catalog No. 502750309
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 120 SFM.
Catalog No. 50-275-0309 Supplier Grainger Supplier No. 14M767
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 120 SFM.