Learn More
Grainger Sq. End Mill Single End Carb 0.0120
Catalog No. 502750345
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling and pocketing in low carbon steels at surface speeds up to 120 SFM when paired with rigid setups.
Catalog No. 50-275-0345 Supplier Grainger Supplier No. 14M755
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling and pocketing in low carbon steels at surface speeds up to 120 SFM when paired with rigid setups.