Learn More
Grainger Sq. End Mill Single End Carb 0.0160
Catalog No. 502750347
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a square end milling cutter made of solid carbide with a TiAlN coating for wear resistance. It has 4 flutes and a 1/2" shank diameter. The overall length is 3" and it has a cutting diameter of 1/2". This is designed for slotting, side milling and contour milling in steels, tool steels, cast irons and other materials. The TiAlN coating provides a smooth finish. This is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-275-0347 Supplier Grainger Supplier No. 14M763
The Micro 100 Square End Mill Carbide is a square end milling cutter made of solid carbide with a TiAlN coating for wear resistance. It has 4 flutes and a 1/2" shank diameter. The overall length is 3" and it has a cutting diameter of 1/2". This is designed for slotting, side milling and contour milling in steels, tool steels, cast irons and other materials. The TiAlN coating provides a smooth finish. This is manufactured in the USA by Micro 100 Tool Corp.