Learn More
Grainger Sq. End Mill Single End Carb 0.0130
Catalog No. 502750348
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. This end mill is uncoated and made in the USA. The operating speed range is 120 to 220 SFM. It can be used on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics.
Catalog No. 50-275-0348 Supplier Grainger Supplier No. 14M756
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. This end mill is uncoated and made in the USA. The operating speed range is 120 to 220 SFM. It can be used on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics.