Learn More
Grainger Sq. End Mill Single End Carb 0.0150
Supplier:  Grainger 14M760
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The end mill has four flutes and a right-hand spiral. It can machine steels, cast iron, aluminum, and other non-ferrous metals at surface speeds up to 120 SFM. The end mill is uncoated and made in the USA.
Catalog No. 50-274-9673