Learn More
Grainger Sq. End Mill Single End Carb 0.0090
Catalog No. 502751999
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. The mill diameter is 0.375 inches with a 0.125 inch depth of cut. This end mill is designed for use on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics. It can be run at speeds up to 2300 SFM.
Catalog No. 50-275-1999 Supplier Grainger Supplier No. 14M748
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. The mill diameter is 0.375 inches with a 0.125 inch depth of cut. This end mill is designed for use on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics. It can be run at speeds up to 2300 SFM.