Learn More
Grainger Sq. End Mill Single End Carb 0.0270
Catalog No. 502752894
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining flat surfaces, slots, and grooves in metal, wood, and plastic. It has a 1/2" shank diameter, 1" overall length, and 1/4" cutting diameter. The carbide tips provide wear resistance and a sharp cutting edge. It has a right-hand spiral flute design for chip ejection. This single end mill is designed for general purpose milling applications.
Catalog No. 50-275-2894 Supplier Grainger Supplier No. 14K512
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining flat surfaces, slots, and grooves in metal, wood, and plastic. It has a 1/2" shank diameter, 1" overall length, and 1/4" cutting diameter. The carbide tips provide wear resistance and a sharp cutting edge. It has a right-hand spiral flute design for chip ejection. This single end mill is designed for general purpose milling applications.