Learn More
Grainger Sq. End Mill Single End Carb 0.0250
Catalog No. 502752931
Encompass_Preferred
The 14K508 is a 14 gauge, 3 conductor SJOOW portable power cable from Southwire. It is rated for 600 volts and designed for hard usage and flexibility in portable power applications. The 3 conductor cable contains a black, white, and green insulated copper wire. It can be used outdoors and in wet areas. The PVC jacket provides durability and is oil resistant. This 50 foot 14 gauge SJOOW cable is an ideal choice for connecting portable generators and distribution equipment.
Catalog No. 50-275-2931 Supplier Grainger Supplier No. 14K508
The 14K508 is a 14 gauge, 3 conductor SJOOW portable power cable from Southwire. It is rated for 600 volts and designed for hard usage and flexibility in portable power applications. The 3 conductor cable contains a black, white, and green insulated copper wire. It can be used outdoors and in wet areas. The PVC jacket provides durability and is oil resistant. This 50 foot 14 gauge SJOOW cable is an ideal choice for connecting portable generators and distribution equipment.