Learn More
Grainger Sq. End Mill Single End Carb 0.90mm
Catalog No. 502753311
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood.
Catalog No. 50-275-3311 Supplier Grainger Supplier No. 14K550
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood.