Learn More
Grainger Sq. End Mill Single End Carb 0.0220
Catalog No. 502751140
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine low carbon steels at surface speeds up to 120 SFM and is designed for use on vertical mills, horizontal mills, and CNC machines. This end mill provides accurate cuts and smooth finishes.
Catalog No. 50-275-1140 Supplier Grainger Supplier No. 14K502
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine low carbon steels at surface speeds up to 120 SFM and is designed for use on vertical mills, horizontal mills, and CNC machines. This end mill provides accurate cuts and smooth finishes.