Learn More
Grainger Sq. End Mill Single End Carb 0.0230
Supplier:  Grainger 14K505
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine aluminum, brass, bronze, cast iron, and plastic at speeds up to 325 SFM. This is a general purpose end mill suitable for a variety of applications.
Catalog No. 50-275-1146