Learn More
Grainger Sq. End Mill Single End Carb 0.70mm
Catalog No. 502751933
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 135 SFM when paired with rigid setups.
Catalog No. 50-275-1933 Supplier Grainger Supplier No. 14K546
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 135 SFM when paired with rigid setups.