Learn More
Grainger Sq. End Mill Single End Carb 0.50mm
Catalog No. 502751514
Encompass_Preferred
The Grainger 14K542 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a satin finish and feature a stamped size on the socket for easy identification. This 108-piece set comes packed in a molded case for storage and portability. The case has a hinged lid and socket rail to keep the sockets organized.
Catalog No. 50-275-1514 Supplier Grainger Supplier No. 14K542
The Grainger 14K542 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a satin finish and feature a stamped size on the socket for easy identification. This 108-piece set comes packed in a molded case for storage and portability. The case has a hinged lid and socket rail to keep the sockets organized.