Learn More
Grainger Sq. End Mill Single End Carb 0.0280
Catalog No. 502751554
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. It can be run at speeds up to 2300 SFM.
Catalog No. 50-275-1554 Supplier Grainger Supplier No. 14K515
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. It can be run at speeds up to 2300 SFM.