Learn More
Grainger Sq. End Mill Single End Carb 0.0160
Catalog No. 502751681
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. This is a general purpose end mill suitable for a variety of applications.
Catalog No. 50-275-1681 Supplier Grainger Supplier No. 14K490
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. This is a general purpose end mill suitable for a variety of applications.