Learn More
Grainger Sq. End Mill Single End Carb 0.0290
Catalog No. 502753187
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for general purpose milling in steel, stainless steel, cast iron, aluminum, brass, and plastics. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The square end measures 1/2 inch across. This is a USA-made end mill suitable for moderate production runs where heat resistance is needed.
Catalog No. 50-275-3187 Supplier Grainger Supplier No. 14K516
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for general purpose milling in steel, stainless steel, cast iron, aluminum, brass, and plastics. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The square end measures 1/2 inch across. This is a USA-made end mill suitable for moderate production runs where heat resistance is needed.