Learn More
Grainger Sq. End Mill Single End Carb 0.0300
Supplier:  Grainger 14K519
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The square end measures 5/16 inch across. This end mill is uncoated and made in the USA. It can machine low carbon steels at surface speeds up to 120 SFM.
Catalog No. 50-275-3362