Learn More
Grainger Sq. End Mill Single End Carb 13/64
Catalog No. 502753209
Encompass_Preferred
The Grainger 14M409 is a 14-inch diameter, 3-blade, aluminum propeller for small unmanned aerial vehicles (UAVs). It has a pitch of 4.7 inches and is designed for electric motors up to 400 Kv. The propeller is made from durable 7075 aluminum alloy and uses a set screw hub attachment. With its lightweight design and optimized blade shape, this propeller provides high thrust while minimizing amp draw. The 14M409 works well for multirotor drones, RC airplanes, and other hobby aircraft needing a 14-inch propeller matched to lower Kv electric motors.
Catalog No. 50-275-3209 Supplier Grainger Supplier No. 14M409
The Grainger 14M409 is a 14-inch diameter, 3-blade, aluminum propeller for small unmanned aerial vehicles (UAVs). It has a pitch of 4.7 inches and is designed for electric motors up to 400 Kv. The propeller is made from durable 7075 aluminum alloy and uses a set screw hub attachment. With its lightweight design and optimized blade shape, this propeller provides high thrust while minimizing amp draw. The 14M409 works well for multirotor drones, RC airplanes, and other hobby aircraft needing a 14-inch propeller matched to lower Kv electric motors.