Learn More
Grainger Sq. End Mill Single End Carb 17/64
Catalog No. 502753321
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill can be used for a variety of materials including low carbon steel, alloy steel, tool steel, stainless steel, cast iron, aluminum, copper alloys, and plastics. It is best suited for light milling applications.
Catalog No. 50-275-3321 Supplier Grainger Supplier No. 14M433
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch corner radius. It is uncoated and made in the USA. The end mill can be used for a variety of materials including low carbon steel, alloy steel, tool steel, stainless steel, cast iron, aluminum, copper alloys, and plastics. It is best suited for light milling applications.