Learn More
Grainger Sq. End Mill Single End Carb 1/2
Catalog No. 502753319
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining flat surfaces, slots, and grooves. It has a 1/2" shank diameter, 1-1/2" overall length, 1/2" cutting diameter, and 2 flute count. The end mill is made of solid carbide for wear resistance and has a TiAIN coating for heat resistance and lubricity. It can machine steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and other non-ferrous materials. This is a high quality carbide end mill for moderate to heavy-duty milling applications.
Catalog No. 50-275-3319 Supplier Grainger Supplier No. 14M516
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining flat surfaces, slots, and grooves. It has a 1/2" shank diameter, 1-1/2" overall length, 1/2" cutting diameter, and 2 flute count. The end mill is made of solid carbide for wear resistance and has a TiAIN coating for heat resistance and lubricity. It can machine steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and other non-ferrous materials. This is a high quality carbide end mill for moderate to heavy-duty milling applications.