Learn More
Grainger Sq. End Mill Single End Carb 11/64
Catalog No. 502753372
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, keyways, pockets, and right angle shoulders. It has a 1/2" shank diameter, 1-1/2" overall length, 1/2" cutting diameter, and 2 flute count. The end mill is made of solid carbide for wear resistance and has a TiAIN coating to reduce friction and heat. It can machine steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and other non-ferrous materials. This durable and precise carbide end mill is well-suited for high production milling applications.
Catalog No. 50-275-3372 Supplier Grainger Supplier No. 14M397
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, keyways, pockets, and right angle shoulders. It has a 1/2" shank diameter, 1-1/2" overall length, 1/2" cutting diameter, and 2 flute count. The end mill is made of solid carbide for wear resistance and has a TiAIN coating to reduce friction and heat. It can machine steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and other non-ferrous materials. This durable and precise carbide end mill is well-suited for high production milling applications.