Learn More
Grainger Sq. End Mill Single End Carb 1/16
Catalog No. 502750175
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM. This end mill is made in the USA.
Catalog No. 50-275-0175 Supplier Grainger Supplier No. 14M354
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM. This end mill is made in the USA.