Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502751330
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and is designed for general purpose milling in most material. The maximum RPM is 2700.
Catalog No. 50-275-1330 Supplier Grainger Supplier No. 14M427
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and is designed for general purpose milling in most material. The maximum RPM is 2700.