Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502749308
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. It is uncoated and made in the USA. This end mill is rigid and versatile for a range of milling applications.
Catalog No. 50-274-9308 Supplier Grainger Supplier No. 14F980
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches. It is uncoated and made in the USA. This end mill is rigid and versatile for a range of milling applications.