Learn More
Grainger Sq. End Mill Single End Carb 1/32
Catalog No. 502749638
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and a 1/2" cutter diameter. The number of flutes is 2. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can slot to a depth of 5/8", and face mill up to 1/2" deep.
Catalog No. 50-274-9638 Supplier Grainger Supplier No. 14T271
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and a 1/2" cutter diameter. The number of flutes is 2. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can slot to a depth of 5/8", and face mill up to 1/2" deep.