Learn More
Grainger Sq. End Mill Single End Carb 1.50mm
Catalog No. 502749759
Encompass_Preferred
The Grainger 14K562 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas auto-darkening filter reacts in 1/25,000th of a second.
Catalog No. 50-274-9759 Supplier Grainger Supplier No. 14K562
The Grainger 14K562 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas auto-darkening filter reacts in 1/25,000th of a second.