Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502749894
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14M251 is a square end milling cutter with a center cutting tip. It has a 1/4" shank diameter and a 1/2" cutting diameter. The overall length is 2-1/2". It is made of solid carbide for wear resistance and has 2 flutes. The square end allows milling square slots and contours. The center cutting tip allows plunging into material. It can be run at speeds up to 325 SFM. This is a versatile square end mill for light milling applications.
Catalog No. 50-274-9894 Supplier Grainger Supplier No. 14M251
The Micro 100 Square End Mill Center Cutting 14M251 is a square end milling cutter with a center cutting tip. It has a 1/4" shank diameter and a 1/2" cutting diameter. The overall length is 2-1/2". It is made of solid carbide for wear resistance and has 2 flutes. The square end allows milling square slots and contours. The center cutting tip allows plunging into material. It can be run at speeds up to 325 SFM. This is a versatile square end mill for light milling applications.